Search
Search
| Gene Symbol: MEP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928416.5 | XP_001928451.2 | 85 | |||
| XM_021098804.1 | XP_020954463.1 | 85 | ||||
| GenBank mRNA | AK345024.1 | - | 5 - 6 | 276 | 85 | |
Set Membership: |
|