Search
Search
| Gene Symbol: RASA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354207.4 | XP_003354255.1 | 764 | 119 | ||
| XM_005661545.3 | XP_005661602.1 | 738 | 119 | |||
| XM_021084515.1 | XP_020940174.1 | 3492 | 119 | |||
| GenBank mRNA | AK345717.1 | - | 2 - 3 | 1256 | 119 | |
Set Membership: |
|