Search
Search
| Gene Symbol: NKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094196.1 | XP_020949855.1 | 413 | 63 | ||
| XM_021094197.1 | XP_020949856.1 | 355 | 63 | |||
| GenBank mRNA | AK345869.1 | - | 3 - 4 | 328 | 63 | |
Set Membership: |
|