Search
Search

SLC6A11| Gene Symbol: SLC6A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069338.1 | XP_020924997.1 | 506 | 109 | ||
| XM_021069339.1 | XP_020924998.1 | 506 | 109 | |||
| GenBank mRNA | AK345949.1 | - | 1 - 2 | 292 | 109 | |
Set Membership: |
|