Search
Search
| Gene Symbol: USP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093724.1 | XP_020949383.1 | 2137 | 77 | ||
| XM_021093725.1 | XP_020949384.1 | 3435 | 77 | |||
| GenBank mRNA | AK346083.1 | - | 5 - 6 | 586 | 77 | |
| AK395883.1 | - | 6 - 7 | 729 | 77 | ||