Search
Search
| Gene Symbol: SBNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073992.1 | XP_020929651.1 | 1565 | 151 | ||
| XM_021073993.1 | XP_020929652.1 | 1559 | 151 | |||
| XM_021073994.1 | XP_020929653.1 | 1190 | 151 | |||
| XM_021073995.1 | XP_020929654.1 | 1188 | 151 | |||
| XM_021073996.1 | XP_020929655.1 | 1469 | 151 | |||
| XM_021073997.1 | XP_020929656.1 | 1160 | 151 | |||
| GenBank mRNA | AK346265.1 | - | 8 - 9 | 1170 | 151 | |
Set Membership: |
|