Search
Search
| Gene Symbol: GNA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005654729.3 | XP_005654786.3 | 4647 | 99 | ||
| GenBank mRNA | AK348023.1 | - | 2 - 3 | 732 | 99 | |
| AK350431.1 | - | 3 - 4 | 476 | 99 | ||
| AK395692.1 | - | 2 - 3 | 736 | 99 | ||
Set Membership: |
|