Search
Search
| Gene Symbol: PIGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928658.5 | XP_001928693.1 | 280 | 90 | ||
| XM_005656345.3 | XP_005656402.1 | 280 | 90 | |||
| XM_005656346.3 | XP_005656403.1 | 280 | 90 | |||
| GenBank mRNA | AK348282.1 | - | 1 - 2 | 233 | 90 | |
Set Membership: |
|