Search
Search
| Gene Symbol: IQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127756.4 | XP_003127804.1 | 396 | 121 | ||
| XM_005665178.3 | XP_005665235.1 | 383 | 121 | |||
| GenBank mRNA | AK349331.1 | - | 2 - 3 | 201 | 121 | |
Set Membership: |
|