Search
Search
| Gene Symbol: EPHB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656047.3 | XP_005656104.2 | 3813 | 118 | ||
| XM_021095499.1 | XP_020951158.1 | 3781 | 118 | |||
| XM_021095500.1 | XP_020951159.1 | 3678 | 118 | |||
| XM_021095501.1 | XP_020951160.1 | 3688 | 118 | |||
| XM_021095502.1 | XP_020951161.1 | 3685 | 118 | |||
| GenBank mRNA | AK349338.1 | - | 1 | 226 | 117 | |
Set Membership: |
|