Search
Search
| Gene Symbol: APLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097131.1 | XP_020952790.1 | 1174 | 65 | ||
| XM_021097132.1 | XP_020952791.1 | 1201 | 65 | |||
| XM_021097133.1 | XP_020952792.1 | 1088 | 65 | |||
| XM_021097134.1 | XP_020952793.1 | 1117 | 65 | |||
| GenBank mRNA | AK349353.1 | - | 8 - 9 | 1127 | 65 | |
| AK391804.1 | - | 8 - 9 | 1110 | 65 | ||
| AK392784.1 | - | 8 - 9 | 1110 | 65 | ||
| AK399912.1 | - | 8 - 9 | 1139 | 65 | ||
Set Membership: |
|