Search
Search

CCDC106| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095055.1 | XP_020950714.1 | 245 | 100 | ||
| XM_021095056.1 | XP_020950715.1 | 285 | 100 | |||
| GenBank mRNA | AK349686.1 | - | 1 - 2 | 285 | 100 | |
Set Membership: |
|