Search
Search
| Gene Symbol: MPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075034.1 | XP_020930693.1 | 1282 | 103 | ||
| XM_021075035.1 | XP_020930694.1 | 1282 | 103 | |||
| GenBank mRNA | AK352226.1 | - | 2 - 3 | 376 | 103 | |
Set Membership: |
|