Search
Search
| Gene Symbol: MPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075034.1 | XP_020930693.1 | 1364 | 82 | ||
| XM_021075035.1 | XP_020930694.1 | 1364 | 82 | |||
| GenBank mRNA | AK352226.1 | - | 3 - 4 | 458 | 82 | |
Set Membership: |
|