Search
Search
| Gene Symbol: RPAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125906.4 | XP_003125954.2 | 313 | 119 | ||
| GenBank mRNA | AK352267.1 | - | 3 - 4 | 249 | 119 | |
| AK391128.1 | - | 3 - 4 | 253 | 119 | ||
Set Membership: |
|