Search
Search
| Gene Symbol: NPHP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995823.2 | XP_013851277.1 | 2161 | 59 | ||
| XM_013995824.2 | XP_013851278.1 | 2158 | 59 | |||
| XM_013995825.2 | XP_013851279.1 | 1975 | 59 | |||
| GenBank mRNA | AK352470.1 | - | 11 - 12 | 1130 | 59 | |
Set Membership: |
|