Search
Search

NDUFA10| Gene Symbol: NDUFA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133785.5 | XP_003133833.1 | 989 | 65 | ||
| GenBank mRNA | AK231733.1 | - | 8 - 9 | 954 | 65 | |
| AK351322.1 | - | 8 - 9 | 921 | 65 | ||
| AK389588.1 | - | 8 - 9 | 976 | 65 | ||
Set Membership: |
|