Search
Search
| Gene Symbol: WDFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065301.1 | XP_020920960.1 | 482 | 71 | ||
| GenBank mRNA | AK231849.1 | - | 1 - 2 | 388 | 71 | |
| AK232818.1 | - | 1 - 2 | 382 | 71 | ||
| AK397418.1 | - | 1 - 2 | 385 | 71 | ||
Set Membership: |
|