Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06902999_m1
See other PBRM1 GE Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
PB1
Chromosome Location:
Chr.13: 34689718 - 34821117 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06902999_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_021068902.1
XP_020924561.1
1032
94
XM_021068903.1
XP_020924562.1
1032
94
XM_021068904.1
XP_020924563.1
1032
94
XM_021068905.1
XP_020924564.1
1032
94
XM_021068906.1
XP_020924565.1
1032
94
XM_021068907.1
XP_020924566.1
1032
94
XM_021068908.1
XP_020924567.1
1032
94
XM_021068909.1
XP_020924568.1
1032
94
XM_021068911.1
XP_020924570.1
1032
94
XM_021068912.1
XP_020924571.1
1032
94
XM_021068913.1
XP_020924572.1
543
94
XM_021068914.1
XP_020924573.1
586
94
XM_021068915.1
XP_020924574.1
484
94
XM_021068916.1
XP_020924575.1
498
94
XM_021068917.1
XP_020924576.1
475
94
XM_021068918.1
XP_020924577.1
543
94
XM_021068919.1
XP_020924578.1
460
94
XM_021068920.1
XP_020924579.1
334
94
XM_021068921.1
XP_020924580.1
449
94
XM_021068922.1
XP_020924581.1
543
94
XM_021068923.1
XP_020924582.1
447
94
XM_021068924.1
XP_020924583.1
429
94
XM_021068925.1
XP_020924584.1
543
94
XM_021068927.1
XP_020924586.1
498
94
XM_021068928.1
XP_020924587.1
543
94
GenBank mRNA
AK231880.1
-
3 - 4
382
94
AK345491.1
-
3 - 4
382
94
AK389997.1
-
4 - 5
543
94
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline