Search
Search
| Gene Symbol: ELK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013990888.2 | XP_013846342.1 | 901 | 97 | ||
| GenBank mRNA | AK232036.1 | - | 2 - 3 | 828 | 97 | |
| AK389972.1 | - | 2 - 3 | 850 | 97 | ||
| AK394179.1 | - | 3 - 4 | 869 | 97 | ||
| AK396610.1 | - | 4 - 5 | 832 | 97 | ||
Set Membership: |
|