Search
Search
| Gene Symbol: MLF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358641.5 | XP_003358689.3 | 752 | 87 | ||
| GenBank mRNA | AK232127.1 | - | 5 - 6 | 664 | 87 | |
| AK399308.1 | - | 5 - 6 | 662 | 87 | ||
Set Membership: |
|