Search
Search

SLC25A44| Gene Symbol: SLC25A44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928054.5 | XP_001928089.1 | 801 | 96 | ||
| XM_013996984.2 | XP_013852438.1 | 917 | 96 | |||
| GenBank mRNA | AK390296.1 | - | 2 - 3 | 778 | 96 | |
Set Membership: |
|