Search
Search
| Gene Symbol: LINGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666134.3 | XP_005666191.1 | 1182 | 69 | ||
| XM_021099021.1 | XP_020954680.1 | 1068 | 69 | |||
| GenBank mRNA | AK390302.1 | - | 2 - 3 | 235 | 69 | |
Set Membership: |
|