Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06905162_m1
See other ANKS1B GE Assays ›
Gene Symbol:
ANKS1B
Gene Name:
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases:
-
Chromosome Location:
Chr.5: 84080505 - 85148742 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06905162_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ANKS1B
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_021092994.1
XP_020948653.1
3632
65
XM_021092995.1
XP_020948654.1
3629
65
XM_021092996.1
XP_020948655.1
3632
65
XM_021092997.1
XP_020948656.1
3629
65
XM_021092998.1
XP_020948657.1
3620
65
XM_021092999.1
XP_020948658.1
3620
65
XM_021093000.1
XP_020948659.1
3911
65
XM_021093001.1
XP_020948660.1
3632
65
XM_021093002.1
XP_020948661.1
3574
65
XM_021093003.1
XP_020948662.1
3545
65
XM_021093004.1
XP_020948663.1
3620
65
XM_021093005.1
XP_020948664.1
3908
65
XM_021093006.1
XP_020948665.1
3530
65
XM_021093007.1
XP_020948666.1
3527
65
XM_021093009.1
XP_020948668.1
3455
65
XM_021093010.1
XP_020948669.1
1684
65
XM_021093011.1
XP_020948670.1
1496
65
XM_021093012.1
XP_020948671.1
1496
65
XM_021093013.1
XP_020948672.1
1204
65
XM_021093014.1
XP_020948673.1
1207
65
XM_021093015.1
XP_020948674.1
1070
65
XM_021093016.1
XP_020948675.1
4117
65
XM_021093017.1
XP_020948676.1
997
65
XM_021093018.1
XP_020948677.1
4034
65
XM_021093020.1
XP_020948679.1
997
65
XM_021093021.1
XP_020948680.1
974
65
GenBank mRNA
AK390352.1
-
11 - 12
1492
65
AK390588.1
-
11 - 12
1492
65
AK400190.1
-
9 - 10
1035
65
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline