Search
Search
| Gene Symbol: EVC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100986.1 | XP_020956645.1 | 481 | 70 | ||
| XM_021100987.1 | XP_020956646.1 | 555 | 70 | |||
| GenBank mRNA | AK390816.1 | - | 1 - 2 | 48 | 70 | |
Set Membership: |
|