Search
Search
| Gene Symbol: FSTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070306.1 | XP_020925965.1 | 1135 | 54 | ||
| GenBank mRNA | AK390819.1 | - | 3 - 4 | 288 | 54 | |
| KF021281.1 | - | 9 - 10 | 867 | 54 | ||
Set Membership: |
|