Search
Search
| Gene Symbol: NRDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099603.1 | XP_020955262.1 | 204 | 103 | ||
| XM_021099604.1 | XP_020955263.1 | 127 | 103 | |||
| GenBank mRNA | AK391031.1 | - | 1 - 2 | 111 | 103 | |
Set Membership: |
|