Search
Search
| Gene Symbol: BBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083234.1 | XP_020938893.1 | 1177 | 105 | ||
| XM_021083235.1 | XP_020938894.1 | 1066 | 105 | |||
| XM_021083236.1 | XP_020938895.1 | 1183 | 105 | |||
| GenBank mRNA | AK391112.1 | - | 3 - 4 | 387 | 105 | |
| AK393528.1 | - | 4 - 5 | 752 | 105 | ||
Set Membership: |
|