Search
Search
| Gene Symbol: CD109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927545.6 | XP_001927580.3 | 2223 | 127 | ||
| XM_021092387.1 | XP_020948046.1 | 1784 | 127 | |||
| GenBank mRNA | AK391289.1 | - | 7 - 8 | 945 | 127 | |
Set Membership: |
|