Search
Search

SLC16A7| Gene Symbol: SLC16A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126337.5 | XP_003126385.1 | 425 | 118 | ||
| XM_013997677.2 | XP_013853131.1 | 233 | 118 | |||
| XM_021091909.1 | XP_020947568.1 | 191 | 118 | |||
| XM_021091910.1 | XP_020947569.1 | 1324 | 118 | |||
| XM_021091911.1 | XP_020947570.1 | 112 | 118 | |||
| GenBank mRNA | AK391347.1 | - | 2 - 3 | 159 | 118 | |
Set Membership: |
|