Search
Search
| Gene Symbol: RPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121581.4 | XP_003121629.2 | 1330 | 72 | ||
| XM_013993150.2 | XP_013848604.2 | 1330 | 72 | |||
| XM_021097423.1 | XP_020953082.1 | 1118 | 72 | |||
| GenBank mRNA | AK391569.1 | - | 9 - 10 | 1288 | 72 | |
Set Membership: |
|