Search
Search
| Gene Symbol: TAGLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070389.1 | XP_020926048.1 | 726 | 93 | ||
| XM_021070390.1 | XP_020926049.1 | 1041 | 93 | |||
| GenBank mRNA | AK391577.1 | - | 4 - 5 | 652 | 93 | |
Set Membership: |
|