Search
Search
| Gene Symbol: PTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926077.6 | XP_001926112.3 | 310 | 113 | ||
| XM_005660513.2 | XP_005660570.2 | 306 | 113 | |||
| XM_021069959.1 | XP_020925618.1 | 305 | 113 | |||
| GenBank mRNA | AK392563.1 | - | 1 - 2 | 217 | 113 | |
Set Membership: |
|