Search
Search
| Gene Symbol: MAU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083430.1 | XP_020939089.1 | 4277 | 76 | ||
| XM_021083431.1 | XP_020939090.1 | 4261 | 76 | |||
| GenBank mRNA | AK393059.1 | - | 1 | 421 | 76 | |
Set Membership: |
|