Search
Search
| Gene Symbol: SENP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069989.1 | XP_020925648.1 | 837 | 81 | ||
| XM_021069990.1 | XP_020925649.1 | 819 | 81 | |||
| XM_021069991.1 | XP_020925650.1 | 835 | 81 | |||
| XM_021069992.1 | XP_020925651.1 | 822 | 81 | |||
| XM_021069993.1 | XP_020925652.1 | 821 | 81 | |||
| XM_021069994.1 | XP_020925653.1 | 913 | 81 | |||
| XM_021069995.1 | XP_020925654.1 | 705 | 81 | |||
| XM_021069996.1 | XP_020925655.1 | 913 | 81 | |||
| GenBank mRNA | AK393245.1 | - | 6 - 7 | 726 | 81 | |
Set Membership: |
|