Search
Search
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357423.4 | XP_003357471.1 | 1111 | 88 | ||
| XM_005667577.3 | XP_005667634.1 | 399 | 88 | |||
| XM_005667578.3 | XP_005667635.1 | 267 | 88 | |||
| GenBank mRNA | AK393692.1 | - | 2 - 3 | 243 | 88 | |
Set Membership: |
|