Search
Search

CARNMT1| Gene Symbol: CARNMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121964.5 | XP_003122012.5 | 814 | 94 | ||
| XM_021064979.1 | XP_020920638.1 | 720 | 94 | |||
| GenBank mRNA | AK393727.1 | - | 3 - 4 | 633 | 94 | |