Search
Search
| Gene Symbol: SLC6A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067520.1 | XP_020923179.1 | 1181 | 98 | ||
| XM_021067521.1 | XP_020923180.1 | 1615 | 98 | |||
| XM_021067522.1 | XP_020923181.1 | 1300 | 98 | |||
| GenBank mRNA | AK393799.1 | - | 1 - 2 | 45 | 98 | |
Set Membership: |
|