Search
Search

SLC39A4| Gene Symbol: SLC39A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925360.5 | XP_001925395.3 | 2551 | 71 | ||
| XM_021090449.1 | XP_020946108.1 | 1743 | 71 | |||
| GenBank mRNA | AK393971.1 | - | 10 - 11 | 1702 | 71 | |
Set Membership: |
|