Search
Search
| Gene Symbol: GIPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083694.1 | XP_020939353.1 | 147 | 77 | ||
| XM_021083695.1 | XP_020939354.1 | 176 | 77 | |||
| XM_021083696.1 | XP_020939355.1 | 162 | 77 | |||
| GenBank mRNA | AK395337.1 | - | 2 - 3 | 124 | 77 | |
Set Membership: |
|