Search
Search
| Gene Symbol: DDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988083.2 | XP_013843537.2 | 957 | 94 | ||
| XM_021089493.1 | XP_020945152.1 | 953 | 94 | |||
| XM_021089494.1 | XP_020945153.1 | 729 | 94 | |||
| XM_021089495.1 | XP_020945154.1 | 1155 | 94 | |||
| XM_021089496.1 | XP_020945155.1 | 982 | 94 | |||
| XM_021089497.1 | XP_020945156.1 | 826 | 94 | |||
| XM_021089498.1 | XP_020945157.1 | 611 | 94 | |||
| XM_021089499.1 | XP_020945158.1 | 786 | 94 | |||
| XM_021089500.1 | XP_020945159.1 | 729 | 94 | |||
| XM_021089501.1 | XP_020945160.1 | 716 | 94 | |||
| GenBank mRNA | AK395607.1 | - | 5 - 6 | 755 | 94 | |
Set Membership: |
|