Search
Search
| Gene Symbol: PRRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090642.1 | XP_020946301.1 | 1652 | 70 | ||
| XM_021090643.1 | XP_020946302.1 | 1651 | 70 | |||
| GenBank mRNA | AK395773.1 | - | 1 - 2 | 681 | 70 | |
Set Membership: |
|