Search
Search
| Gene Symbol: GTF3C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662064.3 | XP_005662121.1 | 294 | 68 | ||
| XM_005662065.3 | XP_005662122.1 | 332 | 68 | |||
| GenBank mRNA | AK395927.1 | - | 1 - 2 | 231 | 68 | |
Set Membership: |
|