Search
Search
| Gene Symbol: HAUS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928749.6 | XP_001928784.1 | 1002 | 104 | ||
| XM_013993527.2 | XP_013848981.1 | 625 | 104 | |||
| GenBank mRNA | AK395981.1 | - | 6 - 7 | 597 | 104 | |
Set Membership: |
|