Search
Search

LOC100625711| Gene Symbol: LOC100625711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359269.4 | XP_003359317.2 | 866 | 97 | ||
| GenBank mRNA | AK396807.1 | - | 5 - 6 | 868 | 97 | |
Set Membership: |
|