Search
Search

KLHDC8B| Gene Symbol: KLHDC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132206.4 | XP_003132254.1 | 1091 | 89 | ||
| XM_005669537.3 | XP_005669594.1 | 1476 | 89 | |||
| XM_013981665.2 | XP_013837119.1 | 1137 | 89 | |||
| XM_013981666.2 | XP_013837120.1 | 929 | 89 | |||
| GenBank mRNA | AK397072.1 | - | 1 - 2 | 92 | 89 | |
Set Membership: |
|