Search
Search

NECTIN4| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013996829.2 | XP_013852283.1 | 1156 | 77 | ||
| XM_013996831.2 | XP_013852285.1 | 1157 | 77 | |||
| XM_021089536.1 | XP_020945195.1 | 1158 | 77 | |||
| XM_021089537.1 | XP_020945196.1 | 1159 | 77 | |||
| XM_021089538.1 | XP_020945197.1 | 1157 | 77 | |||
| GenBank mRNA | AK397273.1 | - | 3 - 4 | 597 | 77 | |
Set Membership: |
|