Search
Search
| Gene Symbol: MMP28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067401.1 | XP_020923060.1 | 1583 | 93 | ||
| XM_021067402.1 | XP_020923061.1 | 1801 | 93 | |||
| XM_021067403.1 | XP_020923062.1 | 1942 | 93 | |||
| GenBank mRNA | AK397377.1 | - | 4 - 5 | 667 | 93 | |
Set Membership: |
|