Search
Search
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077411.1 | XP_020933070.1 | 608 | 113 | ||
| XM_021077412.1 | XP_020933071.1 | 664 | 113 | |||
| GenBank mRNA | AK397720.1 | - | 2 - 3 | 312 | 113 | |
Set Membership: |
|