Search
Search
| Gene Symbol: CMSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981145.2 | XP_013836599.2 | 107 | 78 | ||
| XM_021071314.1 | XP_020926973.1 | 292 | 78 | |||
| XM_021071315.1 | XP_020926974.1 | 316 | 78 | |||
| XM_021071316.1 | XP_020926975.1 | 426 | 78 | |||
| XM_021071317.1 | XP_020926976.1 | 362 | 78 | |||
| GenBank mRNA | AY609865.1 | - | 2 - 3 | 216 | 78 | |
Set Membership: |
|